missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FLNB Partial ORF (NP_001448, 2503 a.a. - 2602 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002317-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: SAIPKASSDASKVTSKGAGLSKAFVGQKSSFLVDCSKAGSNMLLIGVHGPTTPCEEVSMKHVGNQQYNVTYVVKERGDYVLAVKWGEEHIPGSPFHVTVPSpecifications
NP_001448 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SAIPKASSDASKVTSKGAGLSKAFVGQKSSFLVDCSKAGSNMLLIGVHGPTTPCEEVSMKHVGNQQYNVTYVVKERGDYVLAVKWGEEHIPGSPFHVTVP | |
RUO | |
FLNB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2317 | |
FLNB (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABP-278/AOI/DKFZp686A1668/DKFZp686O033/FH1/FLN1L/LRS1/SCT/TABP/TAP | |
FLNB | |
Recombinant | |
wheat germ expression system |