missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FNDC5 Partial ORF (NP_715637.1, 14 a.a. - 78 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00252995-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: SCALWDLEEDTEYIVHVQAISIQGQSPASEPVLFKTPREAEKMASKNKDEVTMKEMGRNQQLRTGSpecifications
NP_715637.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.89kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SCALWDLEEDTEYIVHVQAISIQGQSPASEPVLFKTPREAEKMASKNKDEVTMKEMGRNQQLRTG | |
RUO | |
FNDC5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
252995 | |
FNDC5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FRCP2 | |
FNDC5 | |
Recombinant | |
wheat germ expression system |