missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FOXA3 Partial ORF (NP_004488.2, 266 a.a. - 350 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_004488.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3171 |
Molecular Weight (g/mol) | 35.09kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139444
|
Abnova™
H00003171-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16129444
|
Abnova™
H00003171-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. The crystal structure of a similar protein in rat has been resolved. [provided by RefSeq]
Sequence: EDVGALDCGSPASSTPYFTGLELPGELKLDAPYNFNHPFSINNLMSEQTPAPPKLDVGFGGYGAEGGEPGVYYQGLYSRSLLNASSpecifications
NP_004488.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.09kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKHH3/HNF3G/MGC10179/TCF3G | |
FOXA3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3171 | |
FOXA3 (Human) Recombinant Protein (Q01) | |
EDVGALDCGSPASSTPYFTGLELPGELKLDAPYNFNHPFSINNLMSEQTPAPPKLDVGFGGYGAEGGEPGVYYQGLYSRSLLNAS | |
RUO | |
FOXA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |