missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FOXF2 Partial ORF (NP_001443.1, 346 a.a. - 443 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002295-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138184

  • 6845.00 SEK / 25µg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FOXF2 encodes forkhead box F2, one of many human homologues of the Drosophila melanogaster transcription factor forkhead. FOXF2 is expressed in lung and placenta, and has been shown to transcriptionally activate several lung-specific genes. [provided by RefSeq]

Sequence: SPYLKQPPALTPSSNPAASAGLHSSMSSYSLEQSYLHQNAREDLSVGLPRYQHHSTPVCDRKDFVLNFNGISSFHPSASGSYYHHHHQSVCQDIKPCV
Specifications

Specifications

NP_001443.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue
SPYLKQPPALTPSSNPAASAGLHSSMSSYSLEQSYLHQNAREDLSVGLPRYQHHSTPVCDRKDFVLNFNGISSFHPSASGSYYHHHHQSVCQDIKPCV
RUO
FOXF2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2295
FOXF2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FKHL6/FREAC2
FOXF2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers