missing translation for 'onlineSavingsMsg'
Läs mer

R&D Systems™ Human FoxO1/FKHR (NP_002006) VersaClone cDNA

Produktkod. 18773318
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18773318 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18773318 Leverantör R&D Systems™ Leverantörsnummer RDC1487
  • ONLINE EXCLUSIVE PRICE
    8080.00 SEK
    7641.90 SEK / 10µg
    Save 438.10 SEK
    5% Av
 Begränsad artikel
Beräknad leverans: 26-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Contains gene inserts that are fully sequenced and produced with high quality

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

The Human FoxO1/FKHR (NP_002006) VersaClone cDNA clone contains the complete ORF for FoxO1/FKHR, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The FoxO1/FKHR insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Beställningsinfo

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifikationer

Gen FoxO1/FKHR
Tillträdesnummer NP_002006
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10mM Tris-Cl, pH 8.160
Infoga typ 1981bp
Gen-ID (Entrez) 2308
Gene Alias FKH1, FKHR, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma
Innehåll och lagring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Art Human
Produktlinje R&D Systems
Kvantitet 10 μg
Visa mer Visa mindre

For Reasearch Use Only. Not for use in Humans.

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.