Learn More
Abnova™ Human FPGS Partial ORF (NP_004948, 135 a.a. - 232 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_004948 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2356 |
Molecular Weight (g/mol) | 36.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16148304
|
Abnova™
H00002356-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16138304
|
Abnova™
H00002356-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes the folylpolyglutamate synthetase enzyme. This enzyme has a central role in establishing and maintaining both cytosolic and mitochondrial folylpolyglutamate concentrations and, therefore, is essential for folate homeostasis and the survival of proliferating cells. This enzyme catalyzes the ATP-dependent addition of glutamate moieties to folate and folate derivatives. While several transcript variants may exist for this gene, the full-length natures of only two have been biologically validated to date. These two variants encode distinct isoforms. [provided by RefSeq]
Sequence: QVRERIRINGQPISPELFTKYFWRLYHRLEETKDGSCVSMPPYFRFLTLMAFHVFLQEKVDLAVVEVGIGGAYDCTNIIRKPVVCGVSSLGIDHTSLLSpecifications
NP_004948 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FPGS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2356 | |
FPGS (Human) Recombinant Protein (Q01) | |
QVRERIRINGQPISPELFTKYFWRLYHRLEETKDGSCVSMPPYFRFLTLMAFHVFLQEKVDLAVVEVGIGGAYDCTNIIRKPVVCGVSSLGIDHTSLL | |
RUO | |
FPGS | |
Recombinant | |
wheat germ expression system |