missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FRMPD2 Full-length ORF (NP_001017929.1, 1 a.a. - 261 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_001017929.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 143162 |
Molecular Weight (g/mol) | 55.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16188293
|
Abnova™
H00143162-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16178293
|
Abnova™
H00143162-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a FERM and PDZ domain-containing protein and is located in a region of chromosome 10q which contains a segmental duplication resulting in three nearly identical regions. This copy of the gene is full-length and is in the telomeric duplicated region. The exact function of this gene is unknown; however, FERM domains often play a role in signal transduction pathways. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MGDERTAVSLVTALPGRPSSCVSVTDGPKFEVKLKKNANGLGFSFVQMEKESCSHLKSDLVRIKRLFPGQPAEENGAIAAGDIILAVNGRSTEGLIFQEVLHLLRGAPQEVTLLLCRPPPGALPELEQEWQTPELSADKEFTRATCTDSCTSPILDQEDSWRDSASPDAGEGLGLRPESSQKAIREAQWGQNRERPWASSLTHSPESHPHLCKLHQERDESTLATSLEKDVRQNCYSVCDIMRLGRYSFSSPLTRLSTDIFSpecifications
NP_001017929.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGDERTAVSLVTALPGRPSSCVSVTDGPKFEVKLKKNANGLGFSFVQMEKESCSHLKSDLVRIKRLFPGQPAEENGAIAAGDIILAVNGRSTEGLIFQEVLHLLRGAPQEVTLLLCRPPPGALPELEQEWQTPELSADKEFTRATCTDSCTSPILDQEDSWRDSASPDAGEGLGLRPESSQKAIREAQWGQNRERPWASSLTHSPESHPHLCKLHQERDESTLATSLEKDVRQNCYSVCDIMRLGRYSFSSPLTRLSTDIF | |
RUO | |
FRMPD2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
143162 | |
FRMPD2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC35285/MGC87776/MGC87777/MGC90186/PDZD5C/PDZK5C | |
FRMPD2 | |
Recombinant | |
wheat germ expression system |