missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FRMPD2 Full-length ORF (NP_001017929.1, 1 a.a. - 261 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00143162-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16178293

  • 3343.96 SEK / 10µg

Estimated Shipment: 14-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a FERM and PDZ domain-containing protein and is located in a region of chromosome 10q which contains a segmental duplication resulting in three nearly identical regions. This copy of the gene is full-length and is in the telomeric duplicated region. The exact function of this gene is unknown; however, FERM domains often play a role in signal transduction pathways. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MGDERTAVSLVTALPGRPSSCVSVTDGPKFEVKLKKNANGLGFSFVQMEKESCSHLKSDLVRIKRLFPGQPAEENGAIAAGDIILAVNGRSTEGLIFQEVLHLLRGAPQEVTLLLCRPPPGALPELEQEWQTPELSADKEFTRATCTDSCTSPILDQEDSWRDSASPDAGEGLGLRPESSQKAIREAQWGQNRERPWASSLTHSPESHPHLCKLHQERDESTLATSLEKDVRQNCYSVCDIMRLGRYSFSSPLTRLSTDIF
Specifications

Specifications

NP_001017929.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC35285/MGC87776/MGC87777/MGC90186/PDZD5C/PDZK5C
FRMPD2
Yes
wheat germ expression system
Liquid
143162
FRMPD2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGDERTAVSLVTALPGRPSSCVSVTDGPKFEVKLKKNANGLGFSFVQMEKESCSHLKSDLVRIKRLFPGQPAEENGAIAAGDIILAVNGRSTEGLIFQEVLHLLRGAPQEVTLLLCRPPPGALPELEQEWQTPELSADKEFTRATCTDSCTSPILDQEDSWRDSASPDAGEGLGLRPESSQKAIREAQWGQNRERPWASSLTHSPESHPHLCKLHQERDESTLATSLEKDVRQNCYSVCDIMRLGRYSFSSPLTRLSTDIF
RUO
FRMPD2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates