Learn More
Abnova™ Human FUT4 Partial ORF (NP_002024.1, 167 a.a. - 265 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002526-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The product of this gene transfers fucose to N-acetyllactosamine polysaccharides to generate fucosylated carbohydrate structures. It catalyzes the synthesis of the non-sialylated antigen, Lewis x (CD15). [provided by RefSeq]
Sequence: ITYACWGQLPPLPWASPTPSRPVGVLLWWEPFGGRDSAPRPPPDCRLRFNISGCRLLTDRASYGEAQAVLFHHRDLVKGPPDWPPPWGIQAHTAEEVDLSpecifications
NP_002024.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ITYACWGQLPPLPWASPTPSRPVGVLLWWEPFGGRDSAPRPPPDCRLRFNISGCRLLTDRASYGEAQAVLFHHRDLVKGPPDWPPPWGIQAHTAEEVDL | |
RUO | |
FUT4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2526 | |
FUT4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD15/ELFT/FCT3A/FUC-TIV/FUTIV | |
FUT4 | |
Recombinant | |
wheat germ expression system |