Learn More
Abnova™ Human FZD6 Partial ORF (NP_003497, 71 a.a. - 181 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008323-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene represents a member of the 'frizzled' gene family, which encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The protein encoded by this family member contains a signal peptide, a cysteine-rich domain in the N-terminal extracellular region, and seven transmembrane domains, but unlike other family members, this protein does not contain a C-terminal PDZ domain-binding motif. This protein functions as a negative regulator of the canonical Wnt/beta-catenin signaling cascade, thereby inhibiting the processes that trigger oncogenic transformation, cell proliferation, and inhibition of apoptosis. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: PNIETFLCKAFVPTCIEQIHVVPPCRKLCEKVYSDCKKLIDTFGIRWPEELECDRLQYCDETVPVTFDPHTEFLGPQKKTEQVQRDIGFWCPRHLKTSGGQGYKFLGIDQCSpecifications
NP_003497 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.95kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PNIETFLCKAFVPTCIEQIHVVPPCRKLCEKVYSDCKKLIDTFGIRWPEELECDRLQYCDETVPVTFDPHTEFLGPQKKTEQVQRDIGFWCPRHLKTSGGQGYKFLGIDQC | |
RUO | |
FZD6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8323 | |
FZD6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Hfz6 | |
FZD6 | |
Recombinant | |
wheat germ expression system |