Learn More
Novus Biologicals™ Human Galectin-8 ELISA Kit (Colorimetric)
Human Galectin-8 ELISA Kit (Colorimetric) measures Galectin-8 in Serum, plasma, cell supernatant, and other biological fluids.
Brand:  Novus Biologicals™ NBP3-00386
Product Code. 18321804
  • 7544.08 SEK / 1 set
Explore more special offers
Specifications
0.31 to 20 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Human Galectin-8 in. samples. No significant cross-reactivity or interference between Human Galectin-8 and analogues was observed.
ELISA
LGALS8
Storage of components varies. See protocol for specific instructions.
Galectin-8
0.19 ng/mL
Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Human
3964
1 Kit
Gal-8, galectin 8, galectin-8, galectin-8 g, lectin, galactoside-binding, soluble, 8, PCTA1, PCTA-1 Po66 carbohydrate-binding protein, Po66 carbohydrate binding protein, Po66-CBP, Prostate carcinoma tumor antigen 1