Learn More
Novus Biologicals™ Human Galectin-8 ELISA Kit (Colorimetric)
Human Galectin-8 ELISA Kit (Colorimetric) measures Galectin-8 in Serum, plasma, cell supernatant, and other biological fluids.
Brand: Novus Biologicals™ NBP3-00386
This item is not returnable.
View return policy
Specifications
0.31 to 20 ng/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Human Galectin-8 in. samples. No significant cross-reactivity or interference between Human Galectin-8 and analogues was observed. | |
ELISA | |
LGALS8 | |
Storage of components varies. See protocol for specific instructions. | |
Galectin-8 |
0.19 ng/mL | |
Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum | |
Human | |
3964 | |
1 Kit | |
Gal-8, galectin 8, galectin-8, galectin-8 g, lectin, galactoside-binding, soluble, 8, PCTA1, PCTA-1 Po66 carbohydrate-binding protein, Po66 carbohydrate binding protein, Po66-CBP, Prostate carcinoma tumor antigen 1 |