Learn More
Abnova™ Human GALNT10 Partial ORF (NP_938080, 503 a.a. - 602 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055568-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the polypeptide N-acetylgalactosaminyltransferase (pp-GalNAc-T) gene family. Polypeptide GalNAc transferases initiate the synthesis of mucin-type oligosaccharides by transferring GalNAc from UDP-GalNAc to the hydroxyl group of either a serine or threonine residue on the polypeptide acceptor. Following expression in insect cells, recombinant GalNAc transferase 10 showed significant GalNAcT activity toward mucin-derived peptides, and it utilized both nonglycosylated and glycosylated peptide substrates. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: FTFTWREDIRPGDPQHTKKFCFDAISHTSPVTLYDCHSMKGNQLWKYRKDKTLYHPVSGSCMDCSESDHRIFMNTCNPSSLTQQWLFEHTNSTVLEKFNRSpecifications
NP_938080 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FTFTWREDIRPGDPQHTKKFCFDAISHTSPVTLYDCHSMKGNQLWKYRKDKTLYHPVSGSCMDCSESDHRIFMNTCNPSSLTQQWLFEHTNSTVLEKFNR | |
RUO | |
GALNT10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55568 | |
GALNT10 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586H0623/FLJ00205/FLJ11715/GalNAcT10/pp-GalNAc-T10 | |
GALNT10 | |
Recombinant | |
wheat germ expression system |