missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GALNT13 Partial ORF (NP_443149.1, 47 a.a. - 151 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_443149.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 114805 |
Molecular Weight (g/mol) | 37.29kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16131427
|
Abnova™
H00114805-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16121427
|
Abnova™
H00114805-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The GALNT13 protein is a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAcT; EC 2.4.1.41) family, which initiate O-linked glycosylation of mucins (see MUC3A, MIM 158371) by the initial transfer of N-acetylgalactosamine (GalNAc) with an alpha-linkage to a serine or threonine residue.[supplied by OMIM]
Sequence: RAVISRNQEGPGEMGKAVLIPKDDQEKMKELFKINQFNLMASDLIALNRSLPDVRLEGCKTKVYPDELPNTSVVIVFHNEAWSTLLRTVYSVINRSPHYLLSEVISpecifications
NP_443149.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ16031/FLJ41157/GalNAc-T13/H_NH0187G20.1/KIAA1918/MGC119459/MGC119461/WUGSC:H_NH0187G20.1 | |
GALNT13 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
114805 | |
GALNT13 (Human) Recombinant Protein (Q01) | |
RAVISRNQEGPGEMGKAVLIPKDDQEKMKELFKINQFNLMASDLIALNRSLPDVRLEGCKTKVYPDELPNTSVVIVFHNEAWSTLLRTVYSVINRSPHYLLSEVI | |
RUO | |
GALNT13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |