Learn More
Abnova™ Human GAS2 Full-length ORF (NP_005247.1, 1 a.a. - 313 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002620-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a caspase-3 substrate that plays a role in regulating microfilament and cell shape changes during apoptosis. It can also modulate cell susceptibility to p53-dependent apoptosis by inhibiting calpain activity. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]
Sequence: MCTALSPKVRSGPGLSDMHQYSQWLASRHEANLLPMKEDLALWLTNLLGKEITAETFMEKLDNGALLCQLAETMQEKFKESMDANKPTKNLPLKKIPCKTSAPSGSFFARDNTANFLSWCRDLGVDETCLFESEGLVLHKQPREVCLCLLELGRIAARYGVEPPGLIKLEKEIEQEETLSAPSPSPSPSSKSSGKKSTGNLLDDAVKRISEDPPCKCPNKFCVERLSQGRYRVGEKILFIRMLHNKHVMVRVGGGWETFAGYLLKHDPCRMLQISRVDGKTSPIQSKSPTLKDMNPDNYLVVSASYKAKKEIKSpecifications
NP_005247.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
61.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC32610 | |
GAS2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2620 | |
GAS2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MCTALSPKVRSGPGLSDMHQYSQWLASRHEANLLPMKEDLALWLTNLLGKEITAETFMEKLDNGALLCQLAETMQEKFKESMDANKPTKNLPLKKIPCKTSAPSGSFFARDNTANFLSWCRDLGVDETCLFESEGLVLHKQPREVCLCLLELGRIAARYGVEPPGLIKLEKEIEQEETLSAPSPSPSPSSKSSGKKSTGNLLDDAVKRISEDPPCKCPNKFCVERLSQGRYRVGEKILFIRMLHNKHVMVRVGGGWETFAGYLLKHDPCRMLQISRVDGKTSPIQSKSPTLKDMNPDNYLVVSASYKAKKEIK | |
RUO | |
GAS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |