missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GAS2L1 Full-length ORF (NP_006469.2, 1 a.a. - 681 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_006469.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10634 |
Molecular Weight (g/mol) | 99.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16135462
|
Abnova™
H00010634-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16145462
|
Abnova™
H00010634-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene, a member of the GAS2 family, is similar in sequence to the mouse protein Gas2, an actin-associated protein expressed at high levels in growth-arrested cells. Expression of the mouse Gas2 gene is negatively regulated by serum and growth factors. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MADPVAGIAGSAAKSVRPFRSSEAYVEAMKEDLAEWLNALYGLGLPGGGDGFLTGLATGTTLCQHANAVTEAARALAAARPARGVAFQAHSVVPGSFMARDNVATFIGWCRVELGVPEVLMFETEDLVLRKNEKSVVLCLLEVARRGARLGLLAPRLVQFEQEIERELRAAPPAPNAPAAGEDTTETAPAPGTPARGPRMTPSDLRNLDELVREILGRCTCPDQFPMIKVSEGKYRVGDSSLLIFVRVLRSHVMVRVGGGWDTLEHYLDKHDPCRCSSTAHRPPQPRVCTFSPQRVSPTTSPRPASPVPGSERRGSRPEMTPVSLRSTKEGPETPPRPRDQLPPHPRSRRYSGDSDSSASSAQSGPLGTRSDDTGTGPRRERPSRRLTTGTPASPRRPPALRSQSRDRLDRGRPRGAPGGRGAQLSVPSPARRARSQSREEQAVLLVRRDRDGQHSWVPRGRGSGGSGRSTPQTPRARSPAAPRLSRVSSPSPELGTTPASIFRTPLQLDPQQEQQLFRRLEEEFLANARALEAVASVTPTGPAPDPARAPDPPAPDSAYCSSSSSSSSLSVLGGKCGQPGDSGRTANGLPGPRSQALSSSSDEGSPCPGMGGPLDAPGSPLACTEPSRTWARGRMDTQPDRKPSRIPTPRGPRRPSGPAELGTWHALHSVTPRAEPDSWMSpecifications
NP_006469.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
99.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MADPVAGIAGSAAKSVRPFRSSEAYVEAMKEDLAEWLNALYGLGLPGGGDGFLTGLATGTTLCQHANAVTEAARALAAARPARGVAFQAHSVVPGSFMARDNVATFIGWCRVELGVPEVLMFETEDLVLRKNEKSVVLCLLEVARRGARLGLLAPRLVQFEQEIERELRAAPPAPNAPAAGEDTTETAPAPGTPARGPRMTPSDLRNLDELVREILGRCTCPDQFPMIKVSEGKYRVGDSSLLIFVRVLRSHVMVRVGGGWDTLEHYLDKHDPCRCSSTAHRPPQPRVCTFSPQRVSPTTSPRPASPVPGSERRGSRPEMTPVSLRSTKEGPETPPRPRDQLPPHPRSRRYSGDSDSSASSAQSGPLGTRSDDTGTGPRRERPSRRLTTGTPASPRRPPALRSQSRDRLDRGRPRGAPGGRGAQLSVPSPARRARSQSREEQAVLLVRRDRDGQHSWVPRGRGSGGSGRSTPQTPRARSPAAPRLSRVSSPSPELGTTPASIFRTPLQLDPQQEQQLFRRLEEEFLANARALEAVASVTPTGPAPDPARAPDPPAPDSAYCSSSSSSSSLSVLGGKCGQPGDSGRTANGLPGPRSQALSSSSDEGSPCPGMGGPLDAPGSPLACTEPSRTWARGRMDTQPDRKPSRIPTPRGPRRPSGPAELGTWHALHSVTPRAEPDSWM | |
RUO | |
GAS2L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10634 | |
GAS2L1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GAR22/MGC17243 | |
GAS2L1 | |
Recombinant | |
wheat germ expression system |