missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GATA3 Partial ORF (NP_002042.1, 169 a.a. - 248 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002625-Q01.25ug
Additional Details : Weight : 0.02000kg
Description
Sequence: TPGSAGSARQDEKECLKYQVPLPDSMKLESSHSRGSMTALGGASSSTHHPITTYPPYVPEYSSGLFPPSSLLGGSPTGFGSpecifications
NP_002042.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.54kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
TPGSAGSARQDEKECLKYQVPLPDSMKLESSHSRGSMTALGGASSSTHHPITTYPPYVPEYSSGLFPPSSLLGGSPTGFG | |
RUO | |
GATA3 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
2625 | |
GATA3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HDR|MGC2346|MGC5199|MGC5445 | |
GATA3 | |
Yes |