Learn More
Abnova™ Human GATA4 Partial ORF (NP_002043.2, 343 a.a. - 442 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002626-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in this gene have been associated with cardiac septal defects. [provided by RefSeq]
Sequence: ASGASSNSSNATTSSSEEMRPIKTEPGLSSHYGHSSSVSQTFSVSAMSGHGPSIHPVLSALKLSPQGYASPVSQSPQTSSKQDSWNSLVLADSHGDIITASpecifications
NP_002043.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ASGASSNSSNATTSSSEEMRPIKTEPGLSSHYGHSSSVSQTFSVSAMSGHGPSIHPVLSALKLSPQGYASPVSQSPQTSSKQDSWNSLVLADSHGDIITA | |
RUO | |
GATA4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2626 | |
GATA4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126629 | |
GATA4 | |
Recombinant | |
wheat germ expression system |