missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GATAD2A Partial ORF (NP_060130, 26 a.a. - 134 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054815-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: QIQKEATAQKPTGSVGSTVTTPPPLVRGTQNIPAGKPSLQTSSARMPGSVIPPPLVRGGQQASSKLGPQASSQVVMPPLVRGAQQIHSIRQHSSTGPPPLLLAPRASVPSpecifications
NP_060130 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QIQKEATAQKPTGSVGSTVTTPPPLVRGTQNIPAGKPSLQTSSARMPGSVIPPPLVRGGQQASSKLGPQASSQVVMPPLVRGAQQIHSIRQHSSTGPPPLLLAPRASVP | |
RUO | |
GATAD2A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54815 | |
GATAD2A (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20085/FLJ21017/p66alpha | |
GATAD2A | |
Recombinant | |
wheat germ expression system |