missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GBP6 Full-length ORF (NP_940862.1, 1 a.a. - 633 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4515.00 SEK - 6845.00 SEK

Specifications

Accession Number NP_940862.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 163351
Molecular Weight (g/mol) 98.8kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16128973
View Documents Promotion Details
Abnova™
H00163351-P01.25UG
25 ug
6845.00 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16118973
View Documents Promotion Details
Abnova™
H00163351-P01.10UG
10 ug
4515.00 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Guanylate-binding proteins, such as GBP6, are induced by interferon and hydrolyze GTP to both GDP and GMP (Olszewski et al., 2006 [PubMed 16689661]).[supplied by OMIM]

Sequence: MESGPKMLAPVCLVENNNEQLLVNQQAIQILEKISQPVVVVAIVGLYRTGKSYLMNHLAGQNHGFPLGSTVQSETKGIWMWCVPHPSKPNHTLVLLDTEGLGDVEKGDPKNDSWIFALAVLLCSTFVYNSMSTINHQALEQLHYVTELTELIKAKSSPRPDGVEDSTEFVSFFPDFLWTVRDFTLELKLNGHPITEDEYLENALKLIQGNNPRVQTSNFPRECIRRFFPKRKCFVFDRPTNDKDLLANIEKVSEKQLDPKFQEQTNIFCSYIFTHARTKTLREGITVTGNRLGTLAVTYVEAINSGAVPCLENAVITLAQRENSAAVQRASDYYSQQMAQRVKFPTDTLQELLDVHAACEREAIAIFMEHSFKDENQEFQKKFMETTMNKKGDFLLQNEESSVQYCQAKLNELSKGLMESISAGSFSVPGGHKLYMETKERIEQDYWQVPRKGVKAKEVFQRFLESQMVIEESILQSDKALTDREKAVAVDRAKKEAAEKEQELLKQKLQEQQQQMEAQVKSRKENIAQLKEKLQMEREHLLREQIMMLEHTQKVQNDWLHEGFKKKYEEMNAEISQFKRMIDTTKNDDTPWIARTLDNLADELTAILSAPAKLIGHGVKGVSSLFKKHKLPF
Specifications

Specifications

NP_940862.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
98.8kDa
Glutathione Sepharose 4 Fast Flow
MESGPKMLAPVCLVENNNEQLLVNQQAIQILEKISQPVVVVAIVGLYRTGKSYLMNHLAGQNHGFPLGSTVQSETKGIWMWCVPHPSKPNHTLVLLDTEGLGDVEKGDPKNDSWIFALAVLLCSTFVYNSMSTINHQALEQLHYVTELTELIKAKSSPRPDGVEDSTEFVSFFPDFLWTVRDFTLELKLNGHPITEDEYLENALKLIQGNNPRVQTSNFPRECIRRFFPKRKCFVFDRPTNDKDLLANIEKVSEKQLDPKFQEQTNIFCSYIFTHARTKTLREGITVTGNRLGTLAVTYVEAINSGAVPCLENAVITLAQRENSAAVQRASDYYSQQMAQRVKFPTDTLQELLDVHAACEREAIAIFMEHSFKDENQEFQKKFMETTMNKKGDFLLQNEESSVQYCQAKLNELSKGLMESISAGSFSVPGGHKLYMETKERIEQDYWQVPRKGVKAKEVFQRFLESQMVIEESILQSDKALTDREKAVAVDRAKKEAAEKEQELLKQKLQEQQQQMEAQVKSRKENIAQLKEKLQMEREHLLREQIMMLEHTQKVQNDWLHEGFKKKYEEMNAEISQFKRMIDTTKNDDTPWIARTLDNLADELTAILSAPAKLIGHGVKGVSSLFKKHKLPF
RUO
GBP6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
163351
GBP6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686G0786/FLJ39135
GBP6
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications