Learn More
Abnova™ Human GCLC Full-length ORF (NP_001489.1, 1 a.a. - 637 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002729-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Glutamate-cysteine ligase, also known as gamma-glutamylcysteine synthetase is the first rate limiting enzyme of glutathione synthesis. The enzyme consists of two subunits, a heavy catalytic subunit and a light regulatory subunit. The gene encoding the catalytic subunit encodes a protein of 367 amino acids with a calculated molecular weight of 72.773 kDa and maps to chromosome 6. The regulatory subunit is derived from a different gene located on chromosome 1p22-p21. Deficiency of gamma-glutamylcysteine synthetase in human is associated with enzymopathic hemolytic anemia. [provided by RefSeq]
Sequence: MGLLSQGSPLSWEETKRHADHVRRHGILQFLHIYHAVKDRHKDVLKWGDEVEYMLVSFDHENKKVRLVLSGEKVLETLQEKGERTNPNHPTLWRPEYGSYMIEGTPGQPYGGTMSEFNTVEANMRKRRKEATSILEENQALCTITSFPRLGCPGFTLPEVKPNPVEGGASKSLFFPDEAINKHPRFSTLTRNIRHRRGEKVVINVPIFKDKNTPSPFIETFTEDDEASRASKPDHIYMDAMGFGMGNCCLQVTFQACSISEARYLYDQLATICPIVMALSAASPFYRGYVSDIDCRWGVISASVDDRTREERGLEPLKNNNYRISKSRYDSIDSYLSKCGEKYNDIDLTIDKEIYEQLLQEGIDHLLAQHVAHLFIRDPLTLFEEKIHLDDANESDHFENIQSTNWQTMRFKPPPPNSDIGWRVEFRPMEVQLTDFENSAYVVFVVLLTRVILSYKLDFLIPLSKVDENMKVAQKRDAVLQGMFYFRKDICKGGNAVVDGCGKAQNSTELAAEEYTLMSIDTIINGKEGVFPGLIPILNSYLENMEVDVDTRCSILNYLKLIKKRASGELMTVARWMREFIANHPDYKQDSVITDEMNYSLILKCNQIANELCECPELLGSAFRKVKYSGSKTDSSNSpecifications
NP_001489.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
99.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GCS/GLCL/GLCLC | |
GCLC | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2729 | |
GCLC (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGLLSQGSPLSWEETKRHADHVRRHGILQFLHIYHAVKDRHKDVLKWGDEVEYMLVSFDHENKKVRLVLSGEKVLETLQEKGERTNPNHPTLWRPEYGSYMIEGTPGQPYGGTMSEFNTVEANMRKRRKEATSILEENQALCTITSFPRLGCPGFTLPEVKPNPVEGGASKSLFFPDEAINKHPRFSTLTRNIRHRRGEKVVINVPIFKDKNTPSPFIETFTEDDEASRASKPDHIYMDAMGFGMGNCCLQVTFQACSISEARYLYDQLATICPIVMALSAASPFYRGYVSDIDCRWGVISASVDDRTREERGLEPLKNNNYRISKSRYDSIDSYLSKCGEKYNDIDLTIDKEIYEQLLQEGIDHLLAQHVAHLFIRDPLTLFEEKIHLDDANESDHFENIQSTNWQTMRFKPPPPNSDIGWRVEFRPMEVQLTDFENSAYVVFVVLLTRVILSYKLDFLIPLSKVDENMKVAQKRDAVLQGMFYFRKDICKGGNAVVDGCGKAQNSTELAAEEYTLMSIDTIINGKEGVFPGLIPILNSYLENMEVDVDTRCSILNYLKLIKKRASGELMTVARWMREFIANHPDYKQDSVITDEMNYSLILKCNQIANELCECPELLGSAFRKVKYSGSKTDSSN | |
RUO | |
GCLC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |