Learn More
Abnova™ Human GDF2 Full-length ORF (NP_057288.1, 1 a.a. - 429 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002658-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in rodents suggest that this protein plays a role in the adult liver and in differentiation of cholinergic central nervous system neurons. [provided by RefSeq]
Sequence: MCPGALWVALPLLSLLAGSLQGKPLQSWGRGSAGGNAHSPLGVPGGGLPEHTFNLKMFLENVKVDFLRSLNLSGVPSQDKTRVEPPQYMIDLYNRYTSDKSTTPASNIVRSFSMEDAISITATEDFPFQKHILLFNISIPRHEQITRAELRLYVSCQNHVDPSHDLKGSVVIYDVLDGTDAWDSATETKTFLVSQDIQDEGWETLEVSSAVKRWVRSDSTKSKNKLEVTVESHRKGCDTLDISVPPGSRNLPFFVVFSNDHSSGTKETRLELREMISHEQESVLKKLSKDGSTEAGESSHEEDTDGHVAAGSTLARRKRSAGAGSHCQKTSLRVNFEDIGWDSWIIAPKEYEAYECKGGCFFPLADDVTPTKHAIVQTLVHLKFPTKVGKACCVPTKLSPISVLYKDDMGVPTLKYHYEGMSVAECGCRSpecifications
NP_057288.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BMP-9/BMP9 | |
GDF2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2658 | |
GDF2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MCPGALWVALPLLSLLAGSLQGKPLQSWGRGSAGGNAHSPLGVPGGGLPEHTFNLKMFLENVKVDFLRSLNLSGVPSQDKTRVEPPQYMIDLYNRYTSDKSTTPASNIVRSFSMEDAISITATEDFPFQKHILLFNISIPRHEQITRAELRLYVSCQNHVDPSHDLKGSVVIYDVLDGTDAWDSATETKTFLVSQDIQDEGWETLEVSSAVKRWVRSDSTKSKNKLEVTVESHRKGCDTLDISVPPGSRNLPFFVVFSNDHSSGTKETRLELREMISHEQESVLKKLSKDGSTEAGESSHEEDTDGHVAAGSTLARRKRSAGAGSHCQKTSLRVNFEDIGWDSWIIAPKEYEAYECKGGCFFPLADDVTPTKHAIVQTLVHLKFPTKVGKACCVPTKLSPISVLYKDDMGVPTLKYHYEGMSVAECGCR | |
RUO | |
GDF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |