Learn More
Novus Biologicals™ Human GDNF ELISA Kit (Colorimetric)
Mouse GDNF ELISA Kit (Colorimetric) measures GDNF in Serum, plasma, cell supernatant, and other biological fluids.
Brand:  Novus Biologicals™ NBP3-00392
Product Code. 18323255
  • 7544.08 SEK / 1 set
Explore more special offers
Specifications
0.31 to 20 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Mouse GDNF in samples. No significant cross-reactivity or interference between Mouse GDNF and analogues was observed.
ELISA
GDNF
Apoptosis, Biologically Active Proteins, Cancer
Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3
0.19 ng/mL
Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Mouse
2668
1 Kit
Storage is content dependent.
GDNF