Learn More
Abnova™ Human GDPD1 Full-length ORF (AAH34432.1, 1 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00284161-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the glycerophosphodiester phosphodiesterase family of enzymes that catalyze the hydrolysis of deacylated glycerophospholipids to glycerol phosphate and alcohol. The encoded protein is localized to the cytoplasm and concentrates near the perinuclear region. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MSSTAAFYLLSTLGGYLVTSFLLLKYPTLLHQRKKQRFLSKHISHRGGAGENLENTMAAFQHAVKIGTDMLELDCHITKDEQVVVSHDENLKRATGVNVNISDLKYCELPPYLGKLDVSFQRACQCEGKDNRIPLLKEVFEAFPNTPINIDIKVNNNVLIKKVSELVKRYNREHLTVWGNANYEIVEKCYKENSDIPILFSLQRVLLILGLFFTGLLPFVPIREQFFEIPMPSIILKLKEPHTMSRSQKFLIWLSDLLLMRKALFDHLTARGIQVSFWNDAFWKQHSSPVSpecifications
AAH34432.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ27503/FLJ37451/GDE4/MGC35046/UGPQ | |
GDPD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
284161 | |
GDPD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSSTAAFYLLSTLGGYLVTSFLLLKYPTLLHQRKKQRFLSKHISHRGGAGENLENTMAAFQHAVKIGTDMLELDCHITKDEQVVVSHDENLKRATGVNVNISDLKYCELPPYLGKLDVSFQRACQCEGKDNRIPLLKEVFEAFPNTPINIDIKVNNNVLIKKVSELVKRYNREHLTVWGNANYEIVEKCYKENSDIPILFSLQRVLLILGLFFTGLLPFVPIREQFFEIPMPSIILKLKEPHTMSRSQKFLIWLSDLLLMRKALFDHLTARGIQVSFWNDAFWKQHSSPV | |
RUO | |
GDPD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |