Learn More
Abnova™ Human GFI1 Partial ORF (NP_005254, 1 a.a. - 91 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifica
Accession Number | NP_005254 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2672 |
Molecular Weight (g/mol) | 35.75kDa |
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
16115431
|
Abnova™
H00002672-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16105431
|
Abnova™
H00002672-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descrizione
This gene encodes a nuclear zinc finger protein that functions as a transcriptional repressor. This protein plays a role in diverse developmental contexts, including hematopoiesis and oncogenesis. It functions as part of a complex along with other cofactors to control histone modifications that lead to silencing of the target gene promoters. Mutations in this gene cause autosomal dominant severe congenital neutropenia, and also dominant nonimmune chronic idiopathic neutropenia of adults, which are heterogeneous hematopoietic disorders that cause predispositions to leukemias and infections. Multiple alternatively spliced variants, encoding the same protein, have been identified for this gene. [provided by RefSeq]
Sequence: MPRSFLVKSKKAHSYHQPRSPGPDYSLRLENVPAPSRADSTSNAGGAKAEPRDRLSPESQLTEAPDRASASPRQLRSSVCERSSEFEDFWRSpecifica
NP_005254 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.75kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ94509/GFI-1/ZNF163 | |
GFI1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
2672 | |
GFI1 (Human) Recombinant Protein (Q01) | |
MPRSFLVKSKKAHSYHQPRSPGPDYSLRLENVPAPSRADSTSNAGGAKAEPRDRLSPESQLTEAPDRASASPRQLRSSVCERSSEFEDFWR | |
RUO | |
GFI1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |