Learn More
Abnova™ Human GGT6 Full-length ORF (AAH63111.1, 1 a.a. - 493 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00124975-P01.10ug
Additional Details : Weight : 0.00010kg
Description
GGT6 belongs to the gamma-glutamyltransferase (GGT; EC 2.3.2.2) gene family. GGT is a membrane-bound extracellular enzyme that cleaves gamma-glutamyl peptide bonds in glutathione and other peptides and transfers the gamma-glutamyl moiety to acceptors. GGT is also key to glutathione homeostasis because it provides substrates for glutathione synthesis (Heisterkamp et al., 2008 [PubMed 18357469]).[supplied by OMIM]
Sequence: MERAEEPVVYQKLLPWEPSLESEEEVEEEETSEALVLNPRRHQDSSRNKAGGLPGTWVRVVAALLLLAVGCSLAVRQLQNQGRSTGSLGSVAPPPGGHSHGPGVYHHGAIISPAGRELLVAGGNVVDAGVGAALCLAVVHPHATGLGAMFWGLFHDSSSGNSTALTSGPAQTLAPGLGLPAALPTLHLLHARFGRLPWPRLLVGPTTLAQEGFLVDTPLARALVARGTEGLCPLLCHADGTPLGAGARATNPQLAAVLRSAALAPTSDLAGDALLSLLAGDLGVEVPSAVPRPTLEPAEQLPVPQGILFTTPSPSAGPELLALLEAALRSGAPIPDPCPPFLQTAVSPESSALAAVDSSGSVLLLTSSLNCSFGSAHLSPSTGVLLSNLVAKSTTSAWACPLILRGSLDDTEADVLGLVASGTPDVARAMTHTLLRHLAARPPTQAQHQHQGQQEPTEHPSTCGQGTLLQVAAHTEHAHVSSVPHACCPFQGFSpecifications
AAH63111.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ25990/FLJ90165 | |
GGT6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
124975 | |
GGT6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MERAEEPVVYQKLLPWEPSLESEEEVEEEETSEALVLNPRRHQDSSRNKAGGLPGTWVRVVAALLLLAVGCSLAVRQLQNQGRSTGSLGSVAPPPGGHSHGPGVYHHGAIISPAGRELLVAGGNVVDAGVGAALCLAVVHPHATGLGAMFWGLFHDSSSGNSTALTSGPAQTLAPGLGLPAALPTLHLLHARFGRLPWPRLLVGPTTLAQEGFLVDTPLARALVARGTEGLCPLLCHADGTPLGAGARATNPQLAAVLRSAALAPTSDLAGDALLSLLAGDLGVEVPSAVPRPTLEPAEQLPVPQGILFTTPSPSAGPELLALLEAALRSGAPIPDPCPPFLQTAVSPESSALAAVDSSGSVLLLTSSLNCSFGSAHLSPSTGVLLSNLVAKSTTSAWACPLILRGSLDDTEADVLGLVASGTPDVARAMTHTLLRHLAARPPTQAQHQHQGQQEPTEHPSTCGQGTLLQVAAHTEHAHVSSVPHACCPFQGF | |
RUO | |
GGT6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |