missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GLI1 Partial ORF (AAH13000.1, 401 a.a. - 500 a.a.) Recombinant Protein with GST-pstS1 tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002735-Q02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135491

  • 4515.00 SEK / 10µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the Kruppel family of zinc finger proteins. The encoded transcription factor is activated by the sonic hedgehog signal transduction cascade and regulates stem cell proliferation. The activity and nuclear localization of this protein is negatively regulated by p53 in an inhibitory loop. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: GPLPRAPSISTVEPKREREGGPIREESRLTVPEGAMKPQPSPGAQSSCSSDHSPAGSAANTDSGVEMTGNAGGSTEDLSSLDEGPCIAGTGLSTLRRLEN
Specifications

Specifications

AAH13000.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
39.05kDa
12.5% SDS-PAGE Stained with Coomassie Blue
GPLPRAPSISTVEPKREREGGPIREESRLTVPEGAMKPQPSPGAQSSCSSDHSPAGSAANTDSGVEMTGNAGGSTEDLSSLDEGPCIAGTGLSTLRRLEN
RUO
GLI1
Wheat Germ (in vitro)
GST-pstS1
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2735
GLI1 (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GLI
GLI1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers