Learn More
Novus Biologicals™ Human Glycogen Synthase ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand: Novus Biologicals™ NBP3-11798
This item is not returnable.
View return policy
Specifications
1.25 - 80 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
GYS1 | |
Phospho Specific | |
EC 2.4.1.11, glycogen [starch] synthase, muscle, glycogen synthase 1 (muscle), GYSGSY |
0.75 ng/mL | |
Biological Fluids, Plasma, Serum | |
2997 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Human |