missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GNB5 Partial ORF (NP_057278, 1 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_057278 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10681 |
Molecular Weight (g/mol) | 35.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16163026
|
Abnova™
H00010681-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16153026
|
Abnova™
H00010681-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors. Alternatively spliced transcript variants encoding different isoforms exist. [provided by RefSeq]
Sequence: MCDQTFLVNVFGSCDKCFKQRALRPVFKKSQQLSYCSTCAEIMATEGLHENETLASLKSEAESLKGKLEEERAKLHDVELHQVAERVEALSpecifications
NP_057278 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ37457/FLJ43714/GB5 | |
GNB5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10681 | |
GNB5 (Human) Recombinant Protein (Q01) | |
MCDQTFLVNVFGSCDKCFKQRALRPVFKKSQQLSYCSTCAEIMATEGLHENETLASLKSEAESLKGKLEEERAKLHDVELHQVAERVEAL | |
RUO | |
GNB5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |