missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GNS Partial ORF (NP_002067.1, 455 a.a. - 551 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_002067.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2799 |
Molecular Weight (g/mol) | 36.41kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16014075
|
Abnova™
H00002799-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16004075
|
Abnova™
H00002799-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The product of this gene is a lysosomal enzyme found in all cells. It is involved in the catabolism of heparin, heparan sulphate, and keratan sulphate. Deficiency of this enzyme results in the accumulation of undegraded substrate and the lysosomal storage disorder mucopolysaccharidosis type IIID (Sanfilippo D syndrome). Mucopolysaccharidosis type IIID is the least common of the four subtypes of Sanfilippo syndrome. [provided by RefSeq]
Sequence: VRTMSALWNLQYCEFDDQEVFVEVYNLTADPDQITNIAKTIDPELLGKMNYRLMMLQSCSGPTCRTPGVFDPGYRFDPRLMFSNRGSVRTRRFSKHLSpecifications
NP_002067.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
G6S/MGC21274 | |
GNS | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
2799 | |
GNS (Human) Recombinant Protein (Q01) | |
VRTMSALWNLQYCEFDDQEVFVEVYNLTADPDQITNIAKTIDPELLGKMNYRLMMLQSCSGPTCRTPGVFDPGYRFDPRLMFSNRGSVRTRRFSKHL | |
RUO | |
GNS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |