missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GORASP2 Partial ORF (NP_056345, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4515.00 SEK - 6845.00 SEK

Specifications

Accession Number NP_056345
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 26003
Molecular Weight (g/mol) 36.74kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16175256
View Documents Promotion Details
Abnova™
H00026003-Q01.10UG
10 ug
4515.00 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16185256
View Documents Promotion Details
Abnova™
H00026003-Q01.25UG
25 ug
6845.00 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is involved in establishing the stacked structure of the Golgi apparatus. [provided by RefSeq]

Sequence: MGSSQSVEIPGGGTEGYHVLRVQENSPGHRAGLEPFFDFIVSINGSRLNKDNDTLKDLLKANVEKPVKMLIYSSKTLELRETSVTPSNLWGGQGLLGVSI
Specifications

Specifications

NP_056345
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp434D156/FLJ13139/GOLPH6/GRASP55/GRS2/p59
GORASP2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
26003
GORASP2 (Human) Recombinant Protein (Q01)
MGSSQSVEIPGGGTEGYHVLRVQENSPGHRAGLEPFFDFIVSINGSRLNKDNDTLKDLLKANVEKPVKMLIYSSKTLELRETSVTPSNLWGGQGLLGVSI
RUO
GORASP2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications