Learn More
Abnova™ Human GP9 Full-length ORF (AAH30229.1, 1 a.a. - 177 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002815-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a small membrane glycoprotein found on the surface of human platelets. It forms a 1-to-1 noncovalent complex with glycoprotein Ib, a platelet surface membrane glycoprotein complex that functions as a receptor for von Willebrand factor. The complete receptor complex includes noncovalent association of the alpha and beta subunits with the protein encoded by this gene and platelet glycoprotein V. Defects in this gene are a cause of Bernard-Soulier syndrome, also known as giant platelet disease. These patients have unusually large platelets and have a clinical bleeding tendency. [provided by RefSeq]
Sequence: MPAWGALFLLWATAEATKDCPSPCTCRALETMGLWVDCRGHGLTALPALPARTRHLLLANNSLQSVPPGAFDHLPQLQTLDVTQNPWHCDCSLTYLRLWLEDRTPEALLQVRCASPSLAAHGPLGRLTGYQLGSCGWQLQASWVRPGVLWDVALVTVAALGLALLAGLLCATTEALDSpecifications
AAH30229.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD42a/GPIX | |
GP9 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2815 | |
GP9 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPAWGALFLLWATAEATKDCPSPCTCRALETMGLWVDCRGHGLTALPALPARTRHLLLANNSLQSVPPGAFDHLPQLQTLDVTQNPWHCDCSLTYLRLWLEDRTPEALLQVRCASPSLAAHGPLGRLTGYQLGSCGWQLQASWVRPGVLWDVALVTVAALGLALLAGLLCATTEALD | |
RUO | |
GP9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |