Learn More
Abnova™ Human GPM6B Full-length ORF (NP_001001996.1, 1 a.a. - 305 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002824-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a membrane glycoprotein that belongs to the proteolipid protein family. Proteolipid protein family members are expressed in most brain regions and are thought to be involved in cellular housekeeping functions, such as membrane trafficking and cell-to-cell communication. [provided by RefSeq]
Sequence: MKPAMETAAEENTEQSQERKVNSRAEMEIGRYHWMYPGSKNHQYHPVPTLGDRASPLSSPGCFECCIKCLGGVPYASLVATILCFSGVALFCGCGHVALAGTVAILEQHFSTNASDHALLSEVIQLMQYVIYGIASFFFLYGIILLAEGFYTTSAVKELHGEFKTTACGRCISGMFVFLTYVLGVAWLGVFGFSAVPVFMFYNIWSTCEVIKSPQTNGTTGVEQICVDIRQYGIIPWNAFPGKICGSALENICNTNEFYMSYHLFIVACAGAGATVIALLIYMMATTYNYAVLKFKSREDCCTKFSpecifications
NP_001001996.1 | |
Liquid | |
2824 | |
GPM6B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKPAMETAAEENTEQSQERKVNSRAEMEIGRYHWMYPGSKNHQYHPVPTLGDRASPLSSPGCFECCIKCLGGVPYASLVATILCFSGVALFCGCGHVALAGTVAILEQHFSTNASDHALLSEVIQLMQYVIYGIASFFFLYGIILLAEGFYTTSAVKELHGEFKTTACGRCISGMFVFLTYVLGVAWLGVFGFSAVPVFMFYNIWSTCEVIKSPQTNGTTGVEQICVDIRQYGIIPWNAFPGKICGSALENICNTNEFYMSYHLFIVACAGAGATVIALLIYMMATTYNYAVLKFKSREDCCTKF | |
RUO | |
GPM6B | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
M6B/MGC17150/MGC54284 | |
GPM6B | |
Yes | |
wheat germ expression system |