missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GPSM1 Full-length ORF (AAH09979, 1 a.a. - 434 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH09979 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 26086 |
Molecular Weight (g/mol) | 73.48kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16138062
|
Abnova™
H00026086-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16128062
|
Abnova™
H00026086-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
G proteins propagate intracellular signals initiated by G protein-coupled receptors. GPSM1, a receptor-independent activator of G protein signaling, is one of several factors that influence the basal activity of G protein signaling systems (Pizzinat et al., 2001 [PubMed 11278352]).[supplied by OMIM]
Sequence: MEASCLELALEGERLCKAGDFKTGVAFFEAAVQVGTEDLKTLSAIYSQLGNAYFYLKEHGRALEYHKHDLLLARTIGDRMGEAKASGNLGNTLKVLGRFDEAAVCCQRHLSIAQEQGDKVGEARALYNIGNVYHAKGKQLSWNAANATQDPGHLPPDVRETLCKASEFYERNLSLVKELGDRAAQGRAYGNLGNTHYLLGNFTEATTFHKERLAIAKEFGDKAAERRAYSNLGNAHVFLGRFDVAAEYYKKTLQLSRQLRDQAVEAQACYSLGNTYTLLQDYERAAEYHLRHLLIAQELADRVGEGRACWSLGNAYVSMGRPAQALTFAKKHLQISQEIGDRHGELTARMNVAQLQLVLGRLTSPAASEKPDLAGYEAQGEFQGCGGVLLPTGTDRIRSCGGVGSRPGQHGGGGSRQKMAPTSQFFLASGTAQASpecifications
AAH09979 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.48kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEASCLELALEGERLCKAGDFKTGVAFFEAAVQVGTEDLKTLSAIYSQLGNAYFYLKEHGRALEYHKHDLLLARTIGDRMGEAKASGNLGNTLKVLGRFDEAAVCCQRHLSIAQEQGDKVGEARALYNIGNVYHAKGKQLSWNAANATQDPGHLPPDVRETLCKASEFYERNLSLVKELGDRAAQGRAYGNLGNTHYLLGNFTEATTFHKERLAIAKEFGDKAAERRAYSNLGNAHVFLGRFDVAAEYYKKTLQLSRQLRDQAVEAQACYSLGNTYTLLQDYERAAEYHLRHLLIAQELADRVGEGRACWSLGNAYVSMGRPAQALTFAKKHLQISQEIGDRHGELTARMNVAQLQLVLGRLTSPAASEKPDLAGYEAQGEFQGCGGVLLPTGTDRIRSCGGVGSRPGQHGGGGSRQKMAPTSQFFLASGTAQA | |
RUO | |
GPSM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
26086 | |
GPSM1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AGS3/DKFZp727I051 | |
GPSM1 | |
Recombinant | |
wheat germ expression system |