missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GPSM1 Full-length ORF (AAH09979, 1 a.a. - 434 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026086-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138062

  • 6845.00 SEK / 25µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

G proteins propagate intracellular signals initiated by G protein-coupled receptors. GPSM1, a receptor-independent activator of G protein signaling, is one of several factors that influence the basal activity of G protein signaling systems (Pizzinat et al., 2001 [PubMed 11278352]).[supplied by OMIM]

Sequence: MEASCLELALEGERLCKAGDFKTGVAFFEAAVQVGTEDLKTLSAIYSQLGNAYFYLKEHGRALEYHKHDLLLARTIGDRMGEAKASGNLGNTLKVLGRFDEAAVCCQRHLSIAQEQGDKVGEARALYNIGNVYHAKGKQLSWNAANATQDPGHLPPDVRETLCKASEFYERNLSLVKELGDRAAQGRAYGNLGNTHYLLGNFTEATTFHKERLAIAKEFGDKAAERRAYSNLGNAHVFLGRFDVAAEYYKKTLQLSRQLRDQAVEAQACYSLGNTYTLLQDYERAAEYHLRHLLIAQELADRVGEGRACWSLGNAYVSMGRPAQALTFAKKHLQISQEIGDRHGELTARMNVAQLQLVLGRLTSPAASEKPDLAGYEAQGEFQGCGGVLLPTGTDRIRSCGGVGSRPGQHGGGGSRQKMAPTSQFFLASGTAQA
Specifications

Specifications

AAH09979
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.48kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AGS3/DKFZp727I051
GPSM1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
26086
GPSM1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEASCLELALEGERLCKAGDFKTGVAFFEAAVQVGTEDLKTLSAIYSQLGNAYFYLKEHGRALEYHKHDLLLARTIGDRMGEAKASGNLGNTLKVLGRFDEAAVCCQRHLSIAQEQGDKVGEARALYNIGNVYHAKGKQLSWNAANATQDPGHLPPDVRETLCKASEFYERNLSLVKELGDRAAQGRAYGNLGNTHYLLGNFTEATTFHKERLAIAKEFGDKAAERRAYSNLGNAHVFLGRFDVAAEYYKKTLQLSRQLRDQAVEAQACYSLGNTYTLLQDYERAAEYHLRHLLIAQELADRVGEGRACWSLGNAYVSMGRPAQALTFAKKHLQISQEIGDRHGELTARMNVAQLQLVLGRLTSPAASEKPDLAGYEAQGEFQGCGGVLLPTGTDRIRSCGGVGSRPGQHGGGGSRQKMAPTSQFFLASGTAQA
RUO
GPSM1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers