missing translation for 'onlineSavingsMsg'
Läs mer

Human GRB2 (SH2 Domain) Antibody, R&D Systems™
GREENER_CHOICE

Produktkod. 18761688
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18761688 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18761688 Leverantör R&D Systems Leverantörsnummer MAB38461SP

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody has been used in 2 publications

GRB2 Monoclonal specifically detects GRB2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen GRB2
Användningsområden Western Blot, Immunohistochemistry
Klassificering Monoclonal
Klona 669604
Koncentration LYOPH
Konjugera Unconjugated
Utspädning Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulering Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Genaccessionsnr. P62993
Gene Alias abundant SRC homology, Adapter protein GRB2, ASH, EGFRBP-GRB2, epidermal growth factor receptor-binding protein GRB2, Grb3-3, growth factor receptor-bound protein 2, growth factor receptor-bound protein 3, HT027, MST084, MSTP084, NCKAP2, Protein Ash, SH2/SH3 adapter GRB2
Gensymboler GRB2
Värd art Mouse
Immunogen E. coli-derived recombinant human GRB2 (SH2 Domain) Trp60-Glu152 Accession # P62993
Reningsmetod Protein A or G purified from hybridoma culture supernatant
Kvantitet 25 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 2885
Testspecificitet Detects human GRB2 (SH2 Domain) in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) GRAP2 (SH2 domain; aa 58-149), rhGRB7 (aa 130-274), or rhGRB14 (SH2 domain; aa 439-535) is observed.
Rekonstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Målarter Human
Innehåll och lagring Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotyp IgG2b
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.