missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GRK7 Full-length ORF (AAI52978.1, 1 a.a. - 553 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00131890-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16117953

  • 4515.00 SEK / 10µg
Estimated Shipment: 08-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. It is specifically expressed in the retina and the encoded protein has been shown to phosphorylate cone opsins and initiate their deactivation. [provided by RefSeq]

Sequence: MVDMGALDNLIANTAYLQARKPSDCDSKELQRRRRSLALPGLQGCAELRQKLSLNFHSLCEQQPIGRRLFRDFLATVPTFRKAATFLEDVQNWELAEEGPTKDSALQGLVATCASAPAPGNPQPFLSQAVATKCQAATTEEERVAAVTLAKAEAMAFLQEQPFKDFVTSAFYDKFLQWKLFEMQPVSDKYFTEFRVLGKGGFGEVCAVQVKNTGKMYACKKLDKKRLKKKGGEKMALLEKEILEKVSSPFIVSLAYAFESKTHLCLVMSLMNGGDLKFHIYNVGTRGLDMSRVIFYSAQIACGMLHLHELGIVYRDMKPENVLLDDLGNCRLSDLGLAVEMKGGKPITQRAGTNGYMAPEILMEKVSYSYPVDWFAMGCSIYEMVAGRTPFKDYKEKVSKEDLKQRTLQDEVKFQHDNFTEEAKDICRLFLAKKPEQRLGSREKSDDPRKHHFFKTINFPRLEAGLIEPPFVPDPSVVYAKDIAEIDDFSEVRGVEFDDKDKQFFKNFATGAVPIAWQEEIIETGLFEELNDPNRPTGCEEGNSSKSGVCLLL
Specifications

Specifications

AAI52978.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
87.78kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GPRK7
GRK7
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
131890
GRK7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVDMGALDNLIANTAYLQARKPSDCDSKELQRRRRSLALPGLQGCAELRQKLSLNFHSLCEQQPIGRRLFRDFLATVPTFRKAATFLEDVQNWELAEEGPTKDSALQGLVATCASAPAPGNPQPFLSQAVATKCQAATTEEERVAAVTLAKAEAMAFLQEQPFKDFVTSAFYDKFLQWKLFEMQPVSDKYFTEFRVLGKGGFGEVCAVQVKNTGKMYACKKLDKKRLKKKGGEKMALLEKEILEKVSSPFIVSLAYAFESKTHLCLVMSLMNGGDLKFHIYNVGTRGLDMSRVIFYSAQIACGMLHLHELGIVYRDMKPENVLLDDLGNCRLSDLGLAVEMKGGKPITQRAGTNGYMAPEILMEKVSYSYPVDWFAMGCSIYEMVAGRTPFKDYKEKVSKEDLKQRTLQDEVKFQHDNFTEEAKDICRLFLAKKPEQRLGSREKSDDPRKHHFFKTINFPRLEAGLIEPPFVPDPSVVYAKDIAEIDDFSEVRGVEFDDKDKQFFKNFATGAVPIAWQEEIIETGLFEELNDPNRPTGCEEGNSSKSGVCLLL
RUO
GRK7
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers