Learn More
Abnova™ Human GTF2H5 Full-length ORF (NP_997001.1, 1 a.a. - 71 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00404672-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a subunit of transcription/repair factor TFIIH, which functions in gene transcription and DNA repair. This protein stimulates ERCC3/XPB ATPase activity to trigger DNA opening during DNA repair, and is implicated in regulating cellular levels of TFIIH. Mutations in this gene result in trichothiodystrophy, complementation group A. [provided by RefSeq]
Sequence: MVNVLKGVLIECDPAMKQFLLYLDESNALGKKFIIQDIDDTHVFVIAELVNVLQERVGELMDQNAFSLTQKSpecifications
NP_997001.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C6orf175/TFB5/TGF2H5/TTD/TTD-A/TTDA/bA120J8.2 | |
GTF2H5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
404672 | |
GTF2H5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVNVLKGVLIECDPAMKQFLLYLDESNALGKKFIIQDIDDTHVFVIAELVNVLQERVGELMDQNAFSLTQK | |
RUO | |
GTF2H5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |