missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HABP4 Full-length ORF (AAH18788.1, 1 a.a. - 308 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00022927-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16176752

  • 3343.96 SEK / 10µg

Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MKGALGSPVAAAGAAMQESFGCVVANRFHQLLDDESDPFDILREAERRRQQQLQRKRRDEAAAAAGAGPRGGRSPAGASGHRAGAGGRRESQKERKSLPAPVAQRPDSPGGGLQAPGQKRTPRRGEQQGWNDSRGPEGMLERAERRSYREYRPYETERQADFTAEKFPDEKVPELEVEEETQVQEMTLDEWKNLQEQTRPKPEFNIRKPESTVPSKAVVIHKSKYRDDMVKDDYEDDSHVFRKPANDITSQLEINFGNLPRPGRGARGGTRGGRGRIRRAENYGPRAEVVMQDVAPNPDDPEDFPALS
Specifications

Specifications

AAH18788.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
60.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IHABP4/Ki-1/57/MGC825/SERBP1L
HABP4
Yes
wheat germ expression system
Liquid
22927
HABP4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKGALGSPVAAAGAAMQESFGCVVANRFHQLLDDESDPFDILREAERRRQQQLQRKRRDEAAAAAGAGPRGGRSPAGASGHRAGAGGRRESQKERKSLPAPVAQRPDSPGGGLQAPGQKRTPRRGEQQGWNDSRGPEGMLERAERRSYREYRPYETERQADFTAEKFPDEKVPELEVEEETQVQEMTLDEWKNLQEQTRPKPEFNIRKPESTVPSKAVVIHKSKYRDDMVKDDYEDDSHVFRKPANDITSQLEINFGNLPRPGRGARGGTRGGRGRIRRAENYGPRAEVVMQDVAPNPDDPEDFPALS
RUO
HABP4
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates