Learn More
Abnova™ Human HCFC1 Partial ORF (NP_005325.1, 568 a.a. - 654 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003054-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the host cell factor family and encodes a protein with five Kelch repeats, a fibronectin-like motif, and six HCF repeats, each of which contains a highly specific cleavage signal. This nuclear coactivator is proteolytically cleaved at one of the six possible sites, resulting in the creation of an N-terminal chain and the corresponding C-terminal chain. The final form of this protein consists of noncovalently bound N- and C-terminal chains. The protein is involved in control of the cell cycle and transcriptional regulation during herpes simplex virus infection. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq]
Sequence: KSPISVPGGSALISNLGKVMSVVQTKPVQTSAVTGQASTGPVTQIIQTKGPLPAGTILKLVTSADGKPTTIITTTQASGAGTKPTILSpecifications
NP_005325.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.31kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KSPISVPGGSALISNLGKVMSVVQTKPVQTSAVTGQASTGPVTQIIQTKGPLPAGTILKLVTSADGKPTTIITTTQASGAGTKPTIL | |
RUO | |
HCFC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3054 | |
HCFC1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CFF/HCF-1/HCF1/HFC1/MGC70925/VCAF | |
HCFC1 | |
Recombinant | |
wheat germ expression system |