missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HEL308 Partial ORF (NP_598375, 275 a.a. - 337 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00113510-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121367

  • 6845.00 SEK / 25µg
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

HEL308 is a single-stranded DNA-dependent ATPase and DNA helicase (Marini and Wood, 2002 [PubMed 11751861]).[supplied by OMIM]

Sequence: GNAKAQTPIFSRSKQLKDTLLSEEINVAKKTIESSSNDLGPFYSLPSKVRDLYAQFKGIEKLY
Specifications

Specifications

NP_598375
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.67kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GNAKAQTPIFSRSKQLKDTLLSEEINVAKKTIESSSNDLGPFYSLPSKVRDLYAQFKGIEKLY
RUO
HEL308
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
113510
HEL308 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC20604
HEL308
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers