missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human Helios Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB73091-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Helios Monoclonal specifically detects Helios in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifikationer
| Helios | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 0.5-25 ug/mL | |
| Q9UKS7 | |
| IKZF2 | |
| E. coli-derived recombinant human Helios Thr3-Gln97 Accession # Q9UKS7 | |
| 25 μg | |
| Primary | |
| Detects human Helios in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 736423 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, IKZF2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 22807 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering