missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HERC4 Partial ORF (NP_071362, 341 a.a. - 440 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026091-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138072

  • 6845.00 SEK / 25µg
Estimated Shipment: 02-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

HERC4 belongs to the HERC family of ubiquitin ligases, all of which contain a HECT domain and at least 1 RCC1 (MIM 179710)-like domain (RLD). The 350-amino acid HECT domain is predicted to catalyze the formation of a thioester with ubiquitin before transferring it to a substrate, and the RLD is predicted to act as a guanine nucleotide exchange factor for small G proteins (Hochrainer et al., 2005 [PubMed 15676274]).[supplied by OMIM]

Sequence: VKGNWYPYNGQCLPDIDSEEYFCVKRIFSGGDQSFSHYSSPQNCGPPDDFRCPNPTKQIWTVNEALIQKWLSYPSGRFPVEIANEIDGTFSSSGCLNGSF
Specifications

Specifications

NP_071362
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VKGNWYPYNGQCLPDIDSEEYFCVKRIFSGGDQSFSHYSSPQNCGPPDDFRCPNPTKQIWTVNEALIQKWLSYPSGRFPVEIANEIDGTFSSSGCLNGSF
RUO
HERC4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
26091
HERC4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp564G092/KIAA1593
HERC4
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers