missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HERC4 Partial ORF (NP_071362, 341 a.a. - 440 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_071362 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 26091 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16138072
|
Abnova™
H00026091-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16128072
|
Abnova™
H00026091-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
HERC4 belongs to the HERC family of ubiquitin ligases, all of which contain a HECT domain and at least 1 RCC1 (MIM 179710)-like domain (RLD). The 350-amino acid HECT domain is predicted to catalyze the formation of a thioester with ubiquitin before transferring it to a substrate, and the RLD is predicted to act as a guanine nucleotide exchange factor for small G proteins (Hochrainer et al., 2005 [PubMed 15676274]).[supplied by OMIM]
Sequence: VKGNWYPYNGQCLPDIDSEEYFCVKRIFSGGDQSFSHYSSPQNCGPPDDFRCPNPTKQIWTVNEALIQKWLSYPSGRFPVEIANEIDGTFSSSGCLNGSFSpecifications
NP_071362 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp564G092/KIAA1593 | |
HERC4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
26091 | |
HERC4 (Human) Recombinant Protein (Q01) | |
VKGNWYPYNGQCLPDIDSEEYFCVKRIFSGGDQSFSHYSSPQNCGPPDDFRCPNPTKQIWTVNEALIQKWLSYPSGRFPVEIANEIDGTFSSSGCLNGSF | |
RUO | |
HERC4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |