Learn More
Abnova™ Human HIPK1 Full-length ORF (AAH33012.1, 1 a.a. - 1075 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00204851-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the Ser/Thr family of protein kinases and HIPK subfamily. It phosphorylates homeodomain transcription factors and may also function as a co-repressor for homeodomain transcription factors. Alternative splicing results in four transcript variants encoding four distinct isoforms. [provided by RefSeq]
Sequence: MASQLQVFSPPSVSSSAFCSAKKLKIEPSGWDVSGQSSNDKYYTHSKTLPATQGQANSSHQVANFNIPAYDQGLLLPAPAVEHIVVTAADSSGSAATSTFQSSQTLTHRSNVSLLEPYQKCGLKRKSEEVDSNGSVQIIEEHPPLMLQNRTVVGAAATTTTVTTKSSSSSGEGDYQLVQHEILCSMTNSYEVLEFLGRGTFGQVAKCWKRSTKEIVAIKILKNHPSYARQGQIEVSILSRLSSENADEYNFVRSYECFQHKNHTCLVFEMLEQNLYDFLKQNKFSPLPLKYIRPILQQVATALMKLKSLGLIHADLKPENIMLVDPVRQPYRVKVIDFGSASHVSKAVCSTYLQSRYYRAPEIILGLPFCEAIDMWSLGCVIAELFLGWPLYPGASEYDQIRYISQTQGLPAEYLLSAGTKTTRFFNRDPNLGYPLWRLKTPEEHELETGIKSKEARKYIFNCLDDMAQVNMSTDLEGTDMLAEKADRREYIDLLKKMLTIDADKRITPLKTLNHQFVTMTHLLDFPHSNHVKSCFQNMEICKRRVHMYDTVSQIKSPFTTHVAPNTSTNLTMSFSNQLNTVHNQASVLASSSTAAAATLSLANSDVSLLNYQSALYPSSAAPVPGVAQQGVSLQPGTTQICTQTDPFQQTFIVCPPAFQTGLQATTKHSGFPVRMDNAVPIVPQAPAAQPLQIQSGVLTQGSCTPLMVATLHPQVATITQQYAVPFTLSCAAGRPALVEQTAAVLQAWPGGTQQILLPSTWQQLPGVALHNSVQPTAMIPEAMGSGQQLADWRNAHSHGNQYSTIMQQPSLLTNHVTLATAQPLNVGVAHVVRQQQSSSLPSKKNKQSAPVSSKSSLDVLPSQVYSLVGSSPLRTTSSYNSLVPVQDQHQPIIIPDTPSPPVSVITIRSDTDEEEDNKYKPSSSGLKPRSNVISYVTVNDSPDSDSSLSSPYSTDTLSALRGNSGSVLEGPGRVVADGTGTRTIIVPPLKTQLGDCTVATQASGLLSNKTKPVASVSGQSSGCCITPTGYRAQRGGTSAAQPLNLSQVSAMGYCLLFGPCTVVTFWRTLLLAGCSpecifications
AAH33012.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
143.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0630/MGC26642/MGC33446/MGC33548/Myak/Nbak2 | |
HIPK1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
204851 | |
HIPK1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASQLQVFSPPSVSSSAFCSAKKLKIEPSGWDVSGQSSNDKYYTHSKTLPATQGQANSSHQVANFNIPAYDQGLLLPAPAVEHIVVTAADSSGSAATSTFQSSQTLTHRSNVSLLEPYQKCGLKRKSEEVDSNGSVQIIEEHPPLMLQNRTVVGAAATTTTVTTKSSSSSGEGDYQLVQHEILCSMTNSYEVLEFLGRGTFGQVAKCWKRSTKEIVAIKILKNHPSYARQGQIEVSILSRLSSENADEYNFVRSYECFQHKNHTCLVFEMLEQNLYDFLKQNKFSPLPLKYIRPILQQVATALMKLKSLGLIHADLKPENIMLVDPVRQPYRVKVIDFGSASHVSKAVCSTYLQSRYYRAPEIILGLPFCEAIDMWSLGCVIAELFLGWPLYPGASEYDQIRYISQTQGLPAEYLLSAGTKTTRFFNRDPNLGYPLWRLKTPEEHELETGIKSKEARKYIFNCLDDMAQVNMSTDLEGTDMLAEKADRREYIDLLKKMLTIDADKRITPLKTLNHQFVTMTHLLDFPHSNHVKSCFQNMEICKRRVHMYDTVSQIKSPFTTHVAPNTSTNLTMSFSNQLNTVHNQASVLASSSTAAAATLSLANSDVSLLNYQSALYPSSAAPVPGVAQQGVSLQPGTTQICTQTDPFQQTFIVCPPAFQTGLQATTKHSGFPVRMDNAVPIVPQAPAAQPLQIQSGVLTQGSCTPLMVATLHPQVATITQQYAVPFTLSCAAGRPALVEQTAAVLQAWPGGTQQILLPSTWQQLPGVALHNSVQPTAMIPEAMGSGQQLADWRNAHSHGNQYSTIMQQPSLLTNHVTLATAQPLNVGVAHVVRQQQSSSLPSKKNKQSAPVSSKSSLDVLPSQVYSLVGSSPLRTTSSYNSLVPVQDQHQPIIIPDTPSPPVSVITIRSDTDEEEDNKYKPSSSGLKPRSNVISYVTVNDSPDSDSSLSSPYSTDTLSALRGNSGSVLEGPGRVVADGTGTRTIIVPPLKTQLGDCTVA | |
RUO | |
HIPK1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |