Learn More
Abnova™ Human HIPK2 Partial ORF (AAG41236.1, 961 a.a. - 1065 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00028996-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
HIPK2 is a conserved serine/threonine nuclear kinase that interacts with homeodomain transcription factors.[supplied by OMIM]
Sequence: TGNPRTIIVPPLKTQASEVLVECDSLVPVNTSHHSSSYKSKSSSNVTSTSGHSSGSSSGAITYRQQRPGPHFQQQQPLNLSQAQQHITTDRTGSHRRQQAYITPTSpecifications
AAG41236.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TGNPRTIIVPPLKTQASEVLVECDSLVPVNTSHHSSSYKSKSSSNVTSTSGHSSGSSSGAITYRQQRPGPHFQQQQPLNLSQAQQHITTDRTGSHRRQQAYITPT | |
RUO | |
HIPK2 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
28996 | |
HIPK2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686K02111|FLJ23711|PRO0593 | |
HIPK2 | |
Yes |