missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HNF1B Partial ORF (NP_000449.1, 173 a.a. - 277 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006928-Q02.25ug
Additional Details : Weight : 0.02000kg
Description
Sequence: VRKQREILRQFNQTVQSSGNMTDKSSQDQLLFLFPEFSQQSHGPGQSDDACSEPTNKKMRRNRFKWGPASQQILYQAYDRQKNPSKEEREALVEECNRAECLQRGSpecifications
NP_000449.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
VRKQREILRQFNQTVQSSGNMTDKSSQDQLLFLFPEFSQQSHGPGQSDDACSEPTNKKMRRNRFKWGPASQQILYQAYDRQKNPSKEEREALVEECNRAECLQRG | |
RUO | |
HNF1B | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
6928 | |
HNF1B (Human) Recombinant Protein (Q02) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FJHN|HNF1beta|HNF2|HPC11|LF-B3|LFB3|MODY5|TCF2|VHNF1 | |
HNF1B | |
Yes |