Learn More
Abnova™ Human HOXD3 Partial ORF (NP_008829, 334 a.a. - 431 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_008829 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3232 |
Molecular Weight (g/mol) | 36.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139564
|
Abnova™
H00003232-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16129564
|
Abnova™
H00003232-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that removed the entire HOXD gene cluster or 5' end of this cluster have been associated with severe limb and genital abnormalities. The protein encoded by this gene may play a role in the regulation of cell adhesion processes. [provided by RefSeq]
Sequence: FEPHPMASNGGGFASANLQGSPVYVGGNFVESMAPASGPVFNLGHLSHPSSASVDYSCAAQIPGNHHHGPCDPHPTYTDLSAHHSSQGRLPEAPKLTHSpecifications
NP_008829 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOX1D/HOX4/HOX4A/Hox-4.1/MGC10470 | |
HOXD3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3232 | |
HOXD3 (Human) Recombinant Protein (Q01) | |
FEPHPMASNGGGFASANLQGSPVYVGGNFVESMAPASGPVFNLGHLSHPSSASVDYSCAAQIPGNHHHGPCDPHPTYTDLSAHHSSQGRLPEAPKLTH | |
RUO | |
HOXD3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |