Learn More
Abnova™ Human HPS3 Full-length ORF (AAH16901.1, 1 a.a. - 372 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084343-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein containing a potential clathrin-binding motif, consensus dileucine signals, and tyrosine-based sorting signals for targeting to vesicles of lysosomal lineage. The encoded protein may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 3. Alternate splice variants exist, but their full length sequence has not been determined. [provided by RefSeq]
Sequence: MFYVAEPKQVPHILCSPSMKNINPLTAMSYLRKLDTSGFSSILVTLTKAAVALKMGDLDMHRNEMKSHSEMKLVCGFILEPRLLIQQRKGQIVPTELALHLKETQPGLLVASVLGLQKNNKIGIEEADSFFKVLCAKDEDTIPQLLVDFWEAQLVACLPDVVLQELFFKLTSQYIWRLSKRQPPDTTPLRTSEDLINACSHYGLIYPWVHVVISSDSLADKNYTEDLSKLQSLICGPSFDIASIIPFLEPLSEDTIAGLSVHVLCRTRLKEYEQCIDILLERCPEAVIPYANHELKEENRTLWWKKLLPELCQRIKCGGEKYQLYLSSLKETLSIVAVELELKDFMNVLPEDGTATFFLPYLLYCSRKKPLTSpecifications
AAH16901.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686F0413/FLJ22704/SUTAL | |
HPS3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
84343 | |
HPS3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MFYVAEPKQVPHILCSPSMKNINPLTAMSYLRKLDTSGFSSILVTLTKAAVALKMGDLDMHRNEMKSHSEMKLVCGFILEPRLLIQQRKGQIVPTELALHLKETQPGLLVASVLGLQKNNKIGIEEADSFFKVLCAKDEDTIPQLLVDFWEAQLVACLPDVVLQELFFKLTSQYIWRLSKRQPPDTTPLRTSEDLINACSHYGLIYPWVHVVISSDSLADKNYTEDLSKLQSLICGPSFDIASIIPFLEPLSEDTIAGLSVHVLCRTRLKEYEQCIDILLERCPEAVIPYANHELKEENRTLWWKKLLPELCQRIKCGGEKYQLYLSSLKETLSIVAVELELKDFMNVLPEDGTATFFLPYLLYCSRKKPLT | |
RUO | |
HPS3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |