Learn More
Abnova™ Human HPS6 Partial ORF (NP_079023, 400 a.a. - 500 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079803-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This intronless gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. This protein interacts with Hermansky-Pudlak syndrome 5 protein. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 6. [provided by RefSeq]
Sequence: KDLVFEEACGYYQRRSLRGAQLTPEELRHSSTFRAPQALASILQGHLPPSALLTMLRTELRDYRGLEQLKAQLVAGDDEEAGWTELAEQEVARLLRTELIGSpecifications
NP_079023 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.85kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KDLVFEEACGYYQRRSLRGAQLTPEELRHSSTFRAPQALASILQGHLPPSALLTMLRTELRDYRGLEQLKAQLVAGDDEEAGWTELAEQEVARLLRTELIG | |
RUO | |
HPS6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
79803 | |
HPS6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ22501/MGC20522/RP11-302K17.1 | |
HPS6 | |
Recombinant | |
wheat germ expression system |