Learn More
Abnova™ Human HS6ST1 Partial ORF (NP_004798, 303 a.a. - 401 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_004798 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9394 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16130546
|
Abnova™
H00009394-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16140546
|
Abnova™
H00009394-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the heparan sulfate biosynthetic enzyme family. Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biological activities. This enzyme is a type II integral membrane protein and is responsible for 6-O-sulfation of heparan sulfate. This enzyme does not share significant sequence similarity with other known sulfotransferases. A pseudogene located on chromosome 1 has been found for this gene. [provided by RefSeq]
Sequence: IRPFMQYNSTRAGGVEVDEDTIRRIEELNDLDMQLYDYAKDLFQQRYQYKRQLERREQRLRSREERLLHRAKEALPREDADEPGRVPTEDYMSHIIEKWSpecifications
NP_004798 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp547H098/FLJ25392/HS6ST/MGC116899/MGC116901 | |
HS6ST1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9394 | |
HS6ST1 (Human) Recombinant Protein (Q01) | |
IRPFMQYNSTRAGGVEVDEDTIRRIEELNDLDMQLYDYAKDLFQQRYQYKRQLERREQRLRSREERLLHRAKEALPREDADEPGRVPTEDYMSHIIEKW | |
RUO | |
HS6ST1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |